missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxytyramine Hydrochloride 98.0+%, TCI America™

Catalog No. A03051G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03051G 1 g
A03055G 5 g
A030525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A03051G Supplier TCI America Supplier No. A03051G

Chemical Identifiers

CAS 62-31-7
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.64
MDL Number MFCD00012898
InChI Key CTENFNNZBMHDDG-UHFFFAOYSA-N
Synonym dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril
PubChem CID 65340
IUPAC Name hydrogen 4-(2-aminoethyl)benzene-1,2-diol chloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 189.64
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxytyramine Hydrochloride
EINECSNumber (3)-2870
RTECSNumber UX1092000
TSCA Yes