missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxytyramine Hydrochloride, 99%

Catalog No. p-3736694
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122000100 10 g Glass bottle
AC122001000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122000100 Supplier Thermo Scientific Chemicals Supplier No. 122000100
Only null left

CAS: 62-31-7 | C8H12ClNO2 | 189.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62-31-7
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.64
MDL Number MFCD00012898
InChI Key CTENFNNZBMHDDG-UHFFFAOYSA-N
Synonym dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril
PubChem CID 65340
IUPAC Name 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 241°C to 243°C
Color White
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HO)2C6H3CH2CH2NH2·HCl
Quantity 10 g
Beilstein 13, I, 325
Merck Index 15, 3470
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol and hot 95% ethanol,practically insoluble in ether,chloroform,,benzene and toluene
Formula Weight 189.64
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxytyramine hydrochloride
Show More Show Less
EINECSNumber 200-527-8

RUO – Research Use Only