missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxytetrahydrofuran 97.0+%, TCI America™
$67.38 - $265.67
Chemical Identifiers
| CAS | 453-20-3 |
|---|---|
| Molecular Formula | C4H8O2 |
| Molecular Weight (g/mol) | 88.11 |
| MDL Number | MFCD00005374 |
| InChI Key | XDPCNPCKDGQBAN-UHFFFAOYNA-N |
| Synonym | 3-hydroxytetrahydrofuran, tetrahydrofuran-3-ol, 3-furanol, tetrahydro, tetrahydro-3-furanol, 3-hydroxyoxolane, tetrahydro-furan-3-ol, 3-hydroxytetrahydrofurane, 3-hydroxy tetrahydrofuran, s-3-hydroxytetrahydrofuran, 3-tetrahydrofuranol |
| PubChem CID | 9960 |
| IUPAC Name | oxolan-3-ol |
| SMILES | OC1CCOC1 |
Chemical Identifiers
| 453-20-3 | |
| 88.11 | |
| XDPCNPCKDGQBAN-UHFFFAOYNA-N | |
| 9960 | |
| OC1CCOC1 |
| C4H8O2 | |
| MFCD00005374 | |
| 3-hydroxytetrahydrofuran, tetrahydrofuran-3-ol, 3-furanol, tetrahydro, tetrahydro-3-furanol, 3-hydroxyoxolane, tetrahydro-furan-3-ol, 3-hydroxytetrahydrofurane, 3-hydroxy tetrahydrofuran, s-3-hydroxytetrahydrofuran, 3-tetrahydrofuranol | |
| oxolan-3-ol |
Specifications
| 453-20-3 | |
| 181°C | |
| Clear Liquid at 20°C | |
| MFCD00005374 | |
| XDPCNPCKDGQBAN-UHFFFAOYNA-N | |
| oxolan-3-ol | |
| 9960 | |
| ≥97.0% (GC) |
| Yellow | |
| 5 g | |
| C4H8O2 | |
| 3-hydroxytetrahydrofuran, tetrahydrofuran-3-ol, 3-furanol, tetrahydro, tetrahydro-3-furanol, 3-hydroxyoxolane, tetrahydro-furan-3-ol, 3-hydroxytetrahydrofurane, 3-hydroxy tetrahydrofuran, s-3-hydroxytetrahydrofuran, 3-tetrahydrofuranol | |
| OC1CCOC1 | |
| 88.11 | |
| 88.11 | |
| 3-Hydroxytetrahydrofuran |
Safety and Handling
RTECSNumber : LU3450050
TSCA : Yes