missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxytetrahydrofuran 97.0+%, TCI America™

Catalog No. H13315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13315G 5 g
H133125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13315G Supplier TCI America Supplier No. H13315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 453-20-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00005374
InChI Key XDPCNPCKDGQBAN-UHFFFAOYNA-N
Synonym 3-hydroxytetrahydrofuran, tetrahydrofuran-3-ol, 3-furanol, tetrahydro, tetrahydro-3-furanol, 3-hydroxyoxolane, tetrahydro-furan-3-ol, 3-hydroxytetrahydrofurane, 3-hydroxy tetrahydrofuran, s-3-hydroxytetrahydrofuran, 3-tetrahydrofuranol
PubChem CID 9960
IUPAC Name oxolan-3-ol
SMILES OC1CCOC1

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 181°C
Formula Weight 88.11
Percent Purity ≥97.0% (GC)
Chemical Name or Material 3-Hydroxytetrahydrofuran
RTECSNumber LU3450050
TSCA Yes