missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyquinoline, 97+%, Thermo Scientific™

Chemical Identifiers

CAS 580-18-7
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00169018
InChI Key IQQDNMHUOLMLNJ-UHFFFAOYSA-N
Synonym 3-hydroxyquinoline, 3-quinolinol, 3-quinolol, ccris 4328, unii-12pz582ci0, quinolin-3-o1, 3-hydroxy-quinoline, pubchem12770, 5-21-03-00224 beilstein handbook reference, ksc270k2r
PubChem CID 11376
IUPAC Name quinolin-3-ol
SMILES C1=CC=C2C(=C1)C=C(C=N2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC391372500
SDS View Documents
Thermo Scientific Chemicals
391372500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC391370010
SDS View Documents
Thermo Scientific Chemicals
391370010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

580-18-7
145.16
IQQDNMHUOLMLNJ-UHFFFAOYSA-N
11376
C1=CC=C2C(=C1)C=C(C=N2)O
C9H7NO
MFCD00169018
3-hydroxyquinoline, 3-quinolinol, 3-quinolol, ccris 4328, unii-12pz582ci0, quinolin-3-o1, 3-hydroxy-quinoline, pubchem12770, 5-21-03-00224 beilstein handbook reference, ksc270k2r
quinolin-3-ol

Specifications

580-18-7
100.0
97% min. (HPLC)
C9H7NO
250mg
IQQDNMHUOLMLNJ-UHFFFAOYSA-N
quinolin-3-ol
11376
97+%
97.0
Authentic
Glass bottle
MFCD00169018
3-hydroxyquinoline, 3-quinolinol, 3-quinolol, ccris 4328, unii-12pz582ci0, quinolin-3-o1, 3-hydroxy-quinoline, pubchem12770, 5-21-03-00224 beilstein handbook reference, ksc270k2r
C1=CC=C2C(=C1)C=C(C=N2)O
145.16
145.16
3-Hydroxyquinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-456-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only