missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypyridine N-Oxide 96.0+%, TCI America™
$78.93 - $196.36
Chemical Identifiers
| CAS | 6602-28-4 |
|---|---|
| Molecular Formula | C5H6NO2 |
| Molecular Weight (g/mol) | 112.11 |
| MDL Number | MFCD00006200 |
| InChI Key | MXRXARQXORUYQO-UHFFFAOYNA-N |
| Synonym | 3-hydroxypyridine-n-oxide, 3-hydroxypyridine 1-oxide, 3-hydroxypyridine n-oxide, 3-pyridinol n-oxide, pyridin-3-ol 1-oxide, 3-pyridinol, 1-oxide, 3-hydroxypyridin-1-ium-1-olate, 3-pyridinol-1-oxide, 3-hydroxypyridine-1-oxide, 3-pyridinol 1-oxide |
| PubChem CID | 23069 |
| IUPAC Name | 3-hydroxy-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate |
| SMILES | OC1=CC=C[NH++]([O-])[CH-]1 |
Chemical Identifiers
| 6602-28-4 | |
| 112.11 | |
| MXRXARQXORUYQO-UHFFFAOYNA-N | |
| 23069 | |
| OC1=CC=C[NH++]([O-])[CH-]1 |
| C5H6NO2 | |
| MFCD00006200 | |
| 3-hydroxypyridine-n-oxide, 3-hydroxypyridine 1-oxide, 3-hydroxypyridine n-oxide, 3-pyridinol n-oxide, pyridin-3-ol 1-oxide, 3-pyridinol, 1-oxide, 3-hydroxypyridin-1-ium-1-olate, 3-pyridinol-1-oxide, 3-hydroxypyridine-1-oxide, 3-pyridinol 1-oxide | |
| 3-hydroxy-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate |
Specifications
| 6602-28-4 | |
| C5H6NO2 | |
| 5 g | |
| MXRXARQXORUYQO-UHFFFAOYNA-N | |
| 3-hydroxy-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate | |
| 23069 | |
| ≥96.0% (GC,T) | |
| 3-Hydroxypyridine N-Oxide |
| 190°C | |
| MFCD00006200 | |
| 3-hydroxypyridine-n-oxide, 3-hydroxypyridine 1-oxide, 3-hydroxypyridine n-oxide, 3-pyridinol n-oxide, pyridin-3-ol 1-oxide, 3-pyridinol, 1-oxide, 3-hydroxypyridin-1-ium-1-olate, 3-pyridinol-1-oxide, 3-hydroxypyridine-1-oxide, 3-pyridinol 1-oxide | |
| OC1=CC=C[NH++]([O-])[CH-]1 | |
| 112.11 | |
| 111.10 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UU7719100
TSCA : No