missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyridine-4-carboxylic acid, 98%

Catalog No. p-7054997
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6346503 1 g
AAH6346506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6346503 Supplier Thermo Scientific Chemicals Supplier No. H6346503

CAS: 10128-71-9 | C6H5NO3 | 139.11 g/mol

3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 10128-71-9
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00234165
InChI Key JEHGATQUCUYHJL-UHFFFAOYSA-N
Synonym 3-hydroxyisonicotinic acid, 3-hydroxy-4-pyridinecarboxylic acid, 3-hydroxy-4-pyridinecarboxylicacid, 4-pyridinecarboxylic acid, 3-hydroxy, 3-hydroxy pyridine-4-carboxylic acid, 3-hydroxyisonicontinic acid, pubchem16741, acmc-1bs5o, 3-hydroxy-isonicotinic acid
PubChem CID 459503
IUPAC Name 3-hydroxypyridine-4-carboxylic acid
SMILES C1=CN=CC(=C1C(=O)O)O

Specifications

Quantity 1 g
Beilstein 383677
Solubility Information Soluble in methanol. Slightly soluble in water.
Formula Weight 139.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Hydroxypyridine-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only