missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypropionitrile, 97%

Catalog No. ac149392500
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149392500 250 g Glass bottle
AC149390500 50 g Glass bottle
AC149390010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC149392500 Supplier Thermo Scientific Chemicals Supplier No. 149392500
Only null left

CAS: 109-78-4 | C3H5NO | 71.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-78-4
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00002826
InChI Key WSGYTJNNHPZFKR-UHFFFAOYSA-N
Synonym 3-hydroxypropionitrile, hydracrylonitrile, 2-cyanoethanol, ethylene cyanohydrin, propanenitrile, 3-hydroxy, beta-cyanoethanol, glycol cyanohydrin, methanolacetonitrile, hydroxypropanenitrile, 2-hydroxycyanoethane
PubChem CID 8011
IUPAC Name 3-hydroxypropanenitrile
SMILES C(CO)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -46°C
Color Yellow
Density 1.0400g/mL
Boiling Point 228°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4246 to 1.4266
Linear Formula HOCH2CH2CN
Quantity 250 g
Beilstein 03, 298
Specific Gravity 1.04
Merck Index 15, 3848
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetone,methyl ethyl ketone and,ethyl alcohol,slightly soluble in ether,insoluble in benzene,carbon disulfide and ccl4
Viscosity <10 mPa.s (20°C)
Formula Weight 71.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Hydroxypropionitrile
Show More Show Less
EINECSNumber 203-704-8
RTECSNumber MU5250000
TSCA TSCA

RUO – Research Use Only