missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypiperidine 98.0+%, TCI America™

Catalog No. H04125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04125G 5 g
H041225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04125G Supplier TCI America Supplier No. H04125G

Chemical Identifiers

CAS 6859-99-0
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00014591
InChI Key BIWOSRSKDCZIFM-UHFFFAOYSA-N
Synonym 3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine
PubChem CID 23293
IUPAC Name piperidin-3-ol
SMILES C1CC(CNC1)O

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 101.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxypiperidine