missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypiperidine, 98+%

CAS: 6859-99-0 | C5H11NO | 101.149 g/mol

$37.78 - $342.47

Chemical Identifiers

CAS 6859-99-0
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00014591
InChI Key BIWOSRSKDCZIFM-UHFFFAOYSA-N
Synonym 3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine
PubChem CID 23293
IUPAC Name piperidin-3-ol
SMILES C1CC(CNC1)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0133304
View Documents
Thermo Scientific Chemicals
L0133304
2 g
Each for $37.78
Only null left
 
AAL0133309
View Documents
Thermo Scientific Chemicals
L0133309
10 g
Each for $96.37
Only null left
 
AAL0133318
View Documents
Thermo Scientific Chemicals
L0133318
50 g
Each for $342.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6859-99-0
101.149
BIWOSRSKDCZIFM-UHFFFAOYSA-N
23293
C1CC(CNC1)O
C5H11NO
MFCD00014591
3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine
piperidin-3-ol

Specifications

6859-99-0
11.5 to 11.7
>100°C (212°F)
C5H11NO
2 g
102696
3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine
C1CC(CNC1)O
101.149
101.15
3-Hydroxypiperidine
56°C to 60°C
67°C to 69°C (2 mmHg)
Amine-like
MFCD00014591
UN3263
Air sensitive; Hygroscopic
BIWOSRSKDCZIFM-UHFFFAOYSA-N
piperidin-3-ol
23293
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 229-957-4

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only