missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypiperidine, 98+%

Catalog No. AAL0133304
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0133304 2 g
AAL0133309 10 g
AAL0133318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0133304 Supplier Thermo Scientific Chemicals Supplier No. L0133304
Only null left

CAS: 6859-99-0 | C5H11NO | 101.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6859-99-0
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00014591
InChI Key BIWOSRSKDCZIFM-UHFFFAOYSA-N
Synonym 3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine
PubChem CID 23293
IUPAC Name piperidin-3-ol
SMILES C1CC(CNC1)O

Specifications

Melting Point 56°C to 60°C
pH 11.5 to 11.7
Boiling Point 67°C to 69°C (2 mmHg)
Flash Point >100°C (212°F)
Odor Amine-like
Quantity 2 g
UN Number UN3263
Beilstein 102696
Sensitivity Air sensitive; Hygroscopic
Formula Weight 101.15
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxypiperidine
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 229-957-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only