missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxyphenyl Benzoate 95.0+%, TCI America™
Supplier: TCI America H1560100G
Specifications
| 3-Hydroxyphenyl Benzoate | |
| 135°C | |
| MFCD00020118 | |
| resorcinol monobenzoate, 1,3-benzenediol, monobenzoate, eastman inhibitor rmb, resorcinol, monobenzoate, benzoic acid, m-hydroxyphenyl ester, 3-benzoyloxyphenol, benzoic acid 3-hydroxyphenyl ester, unii-942e82kuwd, 3-hydroxyphenyl benzoate, 1,3-benzenediol, 1-benzoate | |
| OC1=CC=CC(OC(=O)C2=CC=CC=C2)=C1 | |
| 214.22 | |
| 214.22 | |
| Crystalline Powder |
| 136-36-7 | |
| C13H10O3 | |
| 100 g | |
| GDESWOTWNNGOMW-UHFFFAOYSA-N | |
| 3-hydroxyphenyl benzoate | |
| 8690 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 136-36-7 | |
| 214.22 | |
| GDESWOTWNNGOMW-UHFFFAOYSA-N | |
| 8690 | |
| OC1=CC=CC(OC(=O)C2=CC=CC=C2)=C1 |
| C13H10O3 | |
| MFCD00020118 | |
| resorcinol monobenzoate, 1,3-benzenediol, monobenzoate, eastman inhibitor rmb, resorcinol, monobenzoate, benzoic acid, m-hydroxyphenyl ester, 3-benzoyloxyphenol, benzoic acid 3-hydroxyphenyl ester, unii-942e82kuwd, 3-hydroxyphenyl benzoate, 1,3-benzenediol, 1-benzoate | |
| 3-hydroxyphenyl benzoate |
Safety and Handling
EINECSNumber : (3)-1352
RTECSNumber : VH2800000
TSCA : Yes