missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenyl Benzoate 95.0+%, TCI America™

Catalog No. H156025G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H156025G 25 g
H1560100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H156025G Supplier TCI America Supplier No. H156025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 136-36-7
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00020118
InChI Key GDESWOTWNNGOMW-UHFFFAOYSA-N
Synonym resorcinol monobenzoate, 1,3-benzenediol, monobenzoate, eastman inhibitor rmb, resorcinol, monobenzoate, benzoic acid, m-hydroxyphenyl ester, 3-benzoyloxyphenol, benzoic acid 3-hydroxyphenyl ester, unii-942e82kuwd, 3-hydroxyphenyl benzoate, 1,3-benzenediol, 1-benzoate
PubChem CID 8690
IUPAC Name 3-hydroxyphenyl benzoate
SMILES OC1=CC=CC(OC(=O)C2=CC=CC=C2)=C1

Specifications

Melting Point 135°C
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyphenyl Benzoate
EINECSNumber (3)-1352
RTECSNumber VH2800000
TSCA Yes