missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyisoquinoline, 99%

Catalog No. p-7047552
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1942803 1 g
AAL1942806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1942803 Supplier Thermo Scientific Chemicals Supplier No. L1942803
Only null left

CAS: 7651-81-2 | C9H7NO | 145.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7651-81-2
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00075524
InChI Key GYPOFOQUZZUVQL-UHFFFAOYSA-N
Synonym 3-hydroxyisoquinoline, isoquinolin-3-ol, isoquinolin-3 2h-one, 3-isoquinolinol, 2,3-dihydroisoquinolin-3-one, 3 2h-isoquinolinone, 2h-3-isoquinolone, zlchem 163, 3-hydroxy-isoquinoline, 3-isoquinolinol 9ci
PubChem CID 2736554
IUPAC Name 2H-isoquinolin-3-one
SMILES C1=CC2=CC(=O)NC=C2C=C1

Specifications

Melting Point ∽250°C (decomposition)
Quantity 1 g
Beilstein 113230
Formula Weight 145.16
Percent Purity 99%
Chemical Name or Material 3-Hydroxyisoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only