missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyisonicotinic Acid 98.0+%, TCI America™

Catalog No. p-7131013
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13771G 1 g
H13775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13771G Supplier TCI America Supplier No. H13771G

Chemical Identifiers

CAS 10128-71-9
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00234165
InChI Key JEHGATQUCUYHJL-UHFFFAOYSA-N
Synonym 3-hydroxyisonicotinic acid, 3-hydroxy-4-pyridinecarboxylic acid, 3-hydroxy-4-pyridinecarboxylicacid, 4-pyridinecarboxylic acid, 3-hydroxy, 3-hydroxy pyridine-4-carboxylic acid, 3-hydroxyisonicontinic acid, pubchem16741, acmc-1bs5o, 3-hydroxy-isonicotinic acid
PubChem CID 459503
IUPAC Name 3-hydroxypyridine-4-carboxylic acid
SMILES C1=CN=CC(=C1C(=O)O)O

Specifications

Melting Point 315°C
Quantity 1 g
Formula Weight 139.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyisonicotinic Acid