missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxyglutaronitrile 95.0+%, TCI America™
$210.63 - $611.41
Chemical Identifiers
| CAS | 13880-89-2 |
|---|---|
| Molecular Formula | C5H6N2O |
| Molecular Weight (g/mol) | 110.12 |
| MDL Number | MFCD00671545 |
| InChI Key | NMFITULDMUZCQD-UHFFFAOYSA-N |
| Synonym | 1,3-Dicyano-2-propanol |
| PubChem CID | 361762 |
| IUPAC Name | 3-hydroxypentanedinitrile |
| SMILES | OC(CC#N)CC#N |
Chemical Identifiers
| 13880-89-2 | |
| 110.12 | |
| NMFITULDMUZCQD-UHFFFAOYSA-N | |
| 361762 | |
| OC(CC#N)CC#N |
| C5H6N2O | |
| MFCD00671545 | |
| 1,3-Dicyano-2-propanol | |
| 3-hydroxypentanedinitrile |
Specifications
| 13880-89-2 | |
| MFCD00671545 | |
| 3276 | |
| NMFITULDMUZCQD-UHFFFAOYSA-N | |
| 3-hydroxypentanedinitrile | |
| 361762 | |
| ≥95.0% (GC) | |
| 3-Hydroxyglutaronitrile |
| C5H6N2O | |
| 1 g | |
| 1,3-Dicyano-2-propanol | |
| OC(CC#N)CC#N | |
| 110.12 | |
| 110.12 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator