missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyflavone 98.0+%, TCI America™

Catalog No. p-7130516
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03791G 1 g
H037910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03791G Supplier TCI America Supplier No. H03791G
Only null left

Chemical Identifiers

CAS 577-85-5
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.24
MDL Number MFCD00006832
InChI Key HVQAJTFOCKOKIN-UHFFFAOYSA-N
Synonym 3-hydroxyflavone, flavonol, 3-hydroxy-2-phenyl-4h-chromen-4-one, flavon-3-ol, 4h-1-benzopyran-4-one, 3-hydroxy-2-phenyl, flavone, 3-hydroxy, 3-hydroxy-2-phenylchromone, unii-ztg9lss5qh, flavone,3-hydroxy, 3-hf
PubChem CID 11349
ChEBI CHEBI:5078
IUPAC Name 3-hydroxy-2-phenyl-4H-chromen-4-one
SMILES OC1=C(OC2=CC=CC=C2C1=O)C1=CC=CC=C1

Specifications

Melting Point 170°C
Color Yellow
Quantity 1 g
Formula Weight 238.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyflavone
RTECSNumber LK8650000
TSCA Yes