missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyflavone, 98+%

Catalog No. AAA1811003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1811003 1 g
AAA1811006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1811003 Supplier Thermo Scientific Chemicals Supplier No. A1811003

CAS: 577-85-5 | C15H10O3 | 238.24 g/mol

3-Hydroxyflavone is a reactant involved in studies of photochemically-induced dioxygenase-type CO-release reactivity; phase-transfer protection and deprotection of hydroxychromones; and O-methylation with di-Me carbonate. As a reactant it is involved in the synthesis of biologically active molecules including 2-chloropyridine derivatives for studies of antitumor agents and telomerase inhibitors; dihydrochromenopyrazines and chromenoquinoxalines. It is also involved in studies of its electrochemical properties using voltammetric methodologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyflavone is a reactant involved in studies of photochemically-induced dioxygenase-type CO-release reactivity; phase-transfer protection and deprotection of hydroxychromones; and O-methylation with di-Me carbonate. As a reactant it is involved in the synthesis of biologically active molecules including 2-chloropyridine derivatives for studies of antitumor agents and telomerase inhibitors; dihydrochromenopyrazines and chromenoquinoxalines. It is also involved in studies of its electrochemical properties using voltammetric methodologies.

Solubility
Insoluble in water. Soluble in N,N-DMF and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 577-85-5
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.24
MDL Number MFCD00006832
InChI Key HVQAJTFOCKOKIN-UHFFFAOYSA-N
Synonym 3-hydroxyflavone, flavonol, 3-hydroxy-2-phenyl-4h-chromen-4-one, flavon-3-ol, 4h-1-benzopyran-4-one, 3-hydroxy-2-phenyl, flavone, 3-hydroxy, 3-hydroxy-2-phenylchromone, unii-ztg9lss5qh, flavone,3-hydroxy, 3-hf
PubChem CID 11349
ChEBI CHEBI:5078
IUPAC Name 3-hydroxy-2-phenylchromen-4-one
SMILES OC1=C(OC2=CC=CC=C2C1=O)C1=CC=CC=C1

Specifications

Melting Point 168°C to 172°C
Quantity 1 g
Beilstein 15789
Solubility Information Insoluble in water. Soluble in N,N-DMF and ethanol.
Formula Weight 238.25
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxyflavone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 209-416-9
RTECSNumber LK8650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only