missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxycyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. H14081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14081G 1 g
H14085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14081G Supplier TCI America Supplier No. H14081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 606488-94-2
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
InChI Key JBZDHFKPEDWWJC-UHFFFAOYSA-N
PubChem CID 5255726
IUPAC Name 3-hydroxycyclohexane-1-carboxylic acid
SMILES C1CC(CC(C1)O)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 144.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxycyclohexanecarboxylic Acid (cis- and trans- mixture)