missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybutyric acid, 97%

Catalog No. AAH3042606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3042606 5 g
AAH3042614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3042606 Supplier Thermo Scientific Chemicals Supplier No. H3042606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 300-85-6 | C4H8O3 | 104.105 g/mol

3-Hydroxybutyric acid is used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by escherichia coli. It forms films of random copolymer with (R)-3-hydroxypentanoic acid, (R)-3-hydroxyhexanoic acid, 4-hydroxybutyric acid, 6-hydroxyhexanoic acid and (S,S)-lactide by the melt-crystallized method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybutyric acid is used in enantioselective synthesis of D-(-)-3-hydroxybutyric acid by escherichia coli. It forms films of random copolymer with (R)-3-hydroxypentanoic acid, (R)-3-hydroxyhexanoic acid, 4-hydroxybutyric acid, 6-hydroxyhexanoic acid and (S,S)-lactide by the melt-crystallized method.

Solubility
Miscible with water, alcohol and ether.

Notes
Hygroscopic. Incompatible with acids and bases.

Chemical Identifiers

CAS 300-85-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00004546
InChI Key WHBMMWSBFZVSSR-UHFFFAOYSA-N
Synonym 3-hydroxybutyric acid, butanoic acid, 3-hydroxy, beta-hydroxybutyric acid, dl-beta-hydroxybutyric acid, beta-hydroxybuttersaeure, 3 hba, 3-hydroxybuttersaeure, beta-hydroxy-n-butyric acid, 3 hydroxybutyrate, butyric acid, 3-hydroxy
PubChem CID 441
ChEBI CHEBI:20067
IUPAC Name 3-hydroxybutanoic acid
SMILES CC(CC(=O)O)O

Specifications

Melting Point 36°C to 39°C
Odor Odorless
Quantity 5 g
Sensitivity Hygroscopic
Merck Index 14,4816
Solubility Information Miscible with water,alcohol and ether.
Formula Weight 104.1
Percent Purity 98%
Chemical Name or Material 3-Hydroxybutyric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-099-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only