missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzhydrazide, 98+%

Catalog No. p-7044065
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAL0435703 1 g
AAL0435706 5 g
2 options

Catalog No. AAL0435703

Supplier: Thermo Scientific Chemicals L0435703

Only null left

CAS: 5818-06-4 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5818-06-4
MDL Number MFCD00014759

Specifications

Melting Point 157°C to 159°C
Molecular Formula C7H8N2O2
Quantity 1 g
Beilstein 638933
Synonym 3-hydroxybenzhydrazide, 3-hydroxybenzoic hydrazide, m-hydroxybenzohydrazide, m-hydroxybenzhydrazide, 3-hydroxybenzoylhydrazine, benzoylhydrazine, m-hydroxy, 3-???phenol, 3-hydroxy benzhydrazide, 3-hydroxy benzohydrazide, 3-hydroxy-benzohydrazide
InChI Key OACGSLLKFCMXSX-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)O)C(=O)NN
IUPAC Name 3-hydroxybenzohydrazide
Molecular Weight (g/mol) 152.15
PubChem CID 79889
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxybenzhydrazide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-389-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only