missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. H019725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H019725G 25 g
H0197100G 100 g
H0197500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H019725G Supplier TCI America Supplier No. H019725G

Chemical Identifiers

CAS 100-83-4
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00003368
InChI Key IAVREABSGIHHMO-UHFFFAOYSA-N
Synonym m-formylphenol, m-hydroxybenzaldehyde, 3-formylphenol, benzaldehyde, 3-hydroxy, meta-hydroxybenzaldehyde, benzaldehyde, m-hydroxy, 3-hydroxy-benzaldehyde, 3-hydroxy benzaldehyde, 3-hydroxybenzaldehyd, 3-hydroxylbenzaldehyde
PubChem CID 101
ChEBI CHEBI:16207
IUPAC Name 3-hydroxybenzaldehyde
SMILES C1=CC(=CC(=C1)O)C=O

Specifications

Melting Point 104°C
Boiling Point 168°C
Quantity 25 g
Formula Weight 122.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzaldehyde
EINECSNumber (3)-1183&(3)-2660
RTECSNumber CU6450000
TSCA Yes