missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzaldehyde, 98.5%

Catalog No. p-144257
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120961000 100 g Plastic bottle
AC120965000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC120961000 Supplier Thermo Scientific Chemicals Supplier No. 120961000
Only null left

CAS: 100-83-4 | C7H6O2 | 122.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-83-4
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003368
InChI Key IAVREABSGIHHMO-UHFFFAOYSA-N
Synonym m-formylphenol, m-hydroxybenzaldehyde, 3-formylphenol, benzaldehyde, 3-hydroxy, meta-hydroxybenzaldehyde, benzaldehyde, m-hydroxy, 3-hydroxy-benzaldehyde, 3-hydroxy benzaldehyde, 3-hydroxybenzaldehyd, 3-hydroxylbenzaldehyde
PubChem CID 101
ChEBI CHEBI:16207
IUPAC Name 3-hydroxybenzaldehyde
SMILES C1=CC(=CC(=C1)O)C=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 101°C to 105°C
Color Brown to Yellow
Boiling Point 191°C (50.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H4CHO
Quantity 100 g
Beilstein 08, 58
Solubility Information Solubility in water: soluble in water
Formula Weight 122.12
Percent Purity 98.50%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzaldehyde, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 202-892-9
RTECSNumber CU6450000
TSCA TSCA

RUO – Research Use Only