missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzaldehyde, 97%

Catalog No. AAA1354136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1354136 500 g
AAA1354114 25 g
AAA1354122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1354136 Supplier Thermo Scientific Chemicals Supplier No. A1354136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-83-4 | C7H6O2 | 122.123 g/mol

3-Hydroxybenzaldehyde is used as an ionophore, during the development of an highly selective and sensitive PVC membrane sensor, which can be used as a Tb3+ ion selective electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzaldehyde is used as an ionophore, during the development of an highly selective and sensitive PVC membrane sensor, which can be used as a Tb3+ ion selective electrode.

Solubility
Soluble in DMSO and Methanol

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 100-83-4
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00003368
InChI Key IAVREABSGIHHMO-UHFFFAOYSA-N
Synonym m-formylphenol, m-hydroxybenzaldehyde, 3-formylphenol, benzaldehyde, 3-hydroxy, meta-hydroxybenzaldehyde, benzaldehyde, m-hydroxy, 3-hydroxy-benzaldehyde, 3-hydroxy benzaldehyde, 3-hydroxybenzaldehyd, 3-hydroxylbenzaldehyde
PubChem CID 101
ChEBI CHEBI:16207
IUPAC Name 3-hydroxybenzaldehyde
SMILES C1=CC(=CC(=C1)O)C=O

Specifications

Melting Point 102°C to 105°C
Boiling Point 191°C (50 mmHg)
Quantity 500 g
Beilstein 507099
Sensitivity Air Sensitive
Solubility Information Soluble in DMSO and Methanol
Formula Weight 122.12
Percent Purity 97%
Chemical Name or Material 3-Hydroxybenzaldehyde
Hazard Category H302-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 202-892-9
RTECSNumber CU6450000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only