missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyazetidine Hydrochloride 98.0+%, TCI America™

Catalog No. H12641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12641G 1 g
H12645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12641G Supplier TCI America Supplier No. H12641G

Chemical Identifiers

CAS 18621-18-6
Molecular Formula C3H8ClNO
Molecular Weight (g/mol) 109.553
MDL Number MFCD02683887
InChI Key UQUPQEUNHVVNKW-UHFFFAOYSA-N
Synonym 3-hydroxyazetidine hydrochloride, azetidin-3-ol hydrochloride, 3-hydroxyazetidine hcl, azetidin-3-ol hcl, 3-azetidinol hydrochloride, azetidin-3-ol hydrochloride salt, 3-azetidinol, hydrochloride, 3-hydroxy-azetidine hydrochloride, azetidin-3-ol, hydrochloride salt, 3-hydroxy-1-azetidine hydrochloride
PubChem CID 2759290
IUPAC Name azetidin-3-ol;hydrochloride
SMILES C1C(CN1)O.Cl

Specifications

Melting Point 89°C
Quantity 1 g
Formula Weight 109.55
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyazetidine Hydrochloride