missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyazetidine hydrochloride, 97%

Catalog No. AAH2584203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2584203 1 g
AAH2584206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2584203 Supplier Thermo Scientific Chemicals Supplier No. H2584203

CAS: 18621-18-6 | C3H8ClNO | 109.553 g/mol

3-Hydroxyazetidine hydrochloride is an building block used in a recent synthesis of a fluoroquinolone antibiotic and a useful intermediate in the synthesis of polypeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyazetidine hydrochloride is an building block used in a recent synthesis of a fluoroquinolone antibiotic and a useful intermediate in the synthesis of polypeptides.

Solubility
Soluble in water, DMSO, methanol.

Notes
Hygroscopic. Store under inert gas. Store away from water/ moisture, oxidizing agents.

Chemical Identifiers

CAS 18621-18-6
Molecular Formula C3H8ClNO
Molecular Weight (g/mol) 109.553
MDL Number MFCD02683887
InChI Key UQUPQEUNHVVNKW-UHFFFAOYSA-N
Synonym 3-hydroxyazetidine hydrochloride, azetidin-3-ol hydrochloride, 3-hydroxyazetidine hcl, azetidin-3-ol hcl, 3-azetidinol hydrochloride, azetidin-3-ol hydrochloride salt, 3-azetidinol, hydrochloride, 3-hydroxy-azetidine hydrochloride, azetidin-3-ol, hydrochloride salt, 3-hydroxy-1-azetidine hydrochloride
PubChem CID 2759290
IUPAC Name azetidin-3-ol;hydrochloride
SMILES C1C(CN1)O.Cl

Specifications

Melting Point 85°C to 90°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,DMSO,methanol.
Formula Weight 109.56
Percent Purity 97%
Chemical Name or Material 3-Hydroxyazetidine hydrochloride
Hazard Category H302+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only