Learn More
3-Hydroxyazetidine, 95%, Thermo Scientific Chemicals
$135.06 - $452.03
Chemical Identifiers
| CAS | 45347-82-8 |
|---|---|
| Molecular Formula | C3H7NO |
| Molecular Weight (g/mol) | 73.10 |
| MDL Number | MFCD03695446 |
| InChI Key | GMWFCJXSQQHBPI-UHFFFAOYSA-N |
| IUPAC Name | azetidin-3-ol |
| SMILES | OC1CNC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC467210010
|
Thermo Scientific Chemicals
467210010 |
1 g |
Each for $135.06
|
|
|||||
|
AC467210050
|
Thermo Scientific Chemicals
467210050 |
5 g |
Each for $452.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 45347-82-8 | |
| 73.10 | |
| GMWFCJXSQQHBPI-UHFFFAOYSA-N | |
| OC1CNC1 |
| C3H7NO | |
| MFCD03695446 | |
| azetidin-3-ol |
Specifications
| 45347-82-8 | |
| C3H7NO | |
| 1 g | |
| OC1CNC1 | |
| 73.10 | |
| 3-Hydroxyazetidine |
| 95% | |
| MFCD03695446 | |
| GMWFCJXSQQHBPI-UHFFFAOYSA-N | |
| azetidin-3-ol | |
| 73.09 |
Safety and Handling
GHS H Statement Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
RUO – Research Use Only