missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyazetidine, 95%, Thermo Scientific Chemicals

Catalog No. AC467210050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467210050 5 g
AC467210010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467210050 Supplier Thermo Scientific Chemicals Supplier No. 467210050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 45347-82-8
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD03695446
InChI Key GMWFCJXSQQHBPI-UHFFFAOYSA-N
IUPAC Name azetidin-3-ol
SMILES OC1CNC1

Specifications

Assay Percent Range 95%
Quantity 5 g
Formula Weight 73.09
Chemical Name or Material 3-Hydroxyazetidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only