missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyadamantane-1-carboxylic acid, 97%

Catalog No. AAB2037706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2037706 5 g
AAB2037703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2037706 Supplier Thermo Scientific Chemicals Supplier No. B2037706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42711-75-1 | C11H15O3 | 195.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42711-75-1
Molecular Formula C11H15O3
Molecular Weight (g/mol) 195.24
MDL Number MFCD00193929
InChI Key CJJMAWPEZKYJAP-JZVMUCMXSA-M
Synonym 3-hydroxy-1-adamantanecarboxylic acid, 3-hydroxy-adamantane-1-carboxylic acid, 3-hydroxyadamantanecarboxylic acid, 1-carboxy-3-adamantanol, 3-hydroxy-1-adamantane carboxylic acid, 3-hydroxytricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3-carboxy-1-hydroxyadamantane, 3-hydroxy-tricyclo, acmc-1aqmu, 3-carboxy-1-adamantanol
PubChem CID 2736518
IUPAC Name 3-hydroxyadamantane-1-carboxylic acid
SMILES OC12C[C@@H]3C[C@H](C1)CC(C3)(C2)C([O-])=O

Specifications

Melting Point 203°C to 207°C
Quantity 5 g
Formula Weight 196.25
Percent Purity 98%
Chemical Name or Material 3-Hydroxyadamantane-1-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only