missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-6-methyl-2-nitropyridine, 99%

Catalog No. AAL0707418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0707418 50 g
AAL0707409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0707418 Supplier Thermo Scientific Chemicals Supplier No. L0707418

CAS: 15128-90-2 | C6H6N2O3 | 154.125 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15128-90-2
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00006259
InChI Key WZMGQHIBXUAYGS-UHFFFAOYSA-N
PubChem CID 84801
SMILES CC1=NC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 105°C to 108°C
Quantity 50 g
Beilstein 974751
Synonym 3-hydroxy-6-methyl-2-nitropyridine, 6-methyl-2-nitropyridin-3-ol, 6-methyl-2-nitro-3-pyridinol, 3-pyridinol, 6-methyl-2-nitro, 3-hydroxy-2-nitro-6-picoline 3-hydroxy-6-methyl-2-nitropyridine, pubchem6630, 2-nitro-6-methylpyridin-3-ol, 3-pyridinol,6-methyl-2-nitro, 6-methyl-2-nitro-pyridin-3-ol, wzmgqhibxuaygs-uhfffaoysa
Formula Weight 154.13
Percent Purity 99%
Chemical Name or Material 3-Hydroxy-6-methyl-2-nitropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-192-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only