missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-6-methyl-2-nitropyridine 98.0+%, TCI America™
$77.00 - $369.64
Chemical Identifiers
| CAS | 15128-90-2 |
|---|---|
| Molecular Formula | C6H6N2O3 |
| Molecular Weight (g/mol) | 154.125 |
| MDL Number | MFCD00006259 |
| InChI Key | WZMGQHIBXUAYGS-UHFFFAOYSA-N |
| Synonym | 3-hydroxy-6-methyl-2-nitropyridine, 6-methyl-2-nitropyridin-3-ol, 6-methyl-2-nitro-3-pyridinol, 3-pyridinol, 6-methyl-2-nitro, 3-hydroxy-2-nitro-6-picoline 3-hydroxy-6-methyl-2-nitropyridine, pubchem6630, 2-nitro-6-methylpyridin-3-ol, 3-pyridinol,6-methyl-2-nitro, 6-methyl-2-nitro-pyridin-3-ol, wzmgqhibxuaygs-uhfffaoysa |
| PubChem CID | 84801 |
| SMILES | CC1=NC(=C(C=C1)O)[N+](=O)[O-] |
Chemical Identifiers
| 15128-90-2 | |
| 154.125 | |
| WZMGQHIBXUAYGS-UHFFFAOYSA-N | |
| 84801 |
| C6H6N2O3 | |
| MFCD00006259 | |
| 3-hydroxy-6-methyl-2-nitropyridine, 6-methyl-2-nitropyridin-3-ol, 6-methyl-2-nitro-3-pyridinol, 3-pyridinol, 6-methyl-2-nitro, 3-hydroxy-2-nitro-6-picoline 3-hydroxy-6-methyl-2-nitropyridine, pubchem6630, 2-nitro-6-methylpyridin-3-ol, 3-pyridinol,6-methyl-2-nitro, 6-methyl-2-nitro-pyridin-3-ol, wzmgqhibxuaygs-uhfffaoysa | |
| CC1=NC(=C(C=C1)O)[N+](=O)[O-] |
Specifications
| 15128-90-2 | |
| Yellow | |
| MFCD00006259 | |
| 3-hydroxy-6-methyl-2-nitropyridine, 6-methyl-2-nitropyridin-3-ol, 6-methyl-2-nitro-3-pyridinol, 3-pyridinol, 6-methyl-2-nitro, 3-hydroxy-2-nitro-6-picoline 3-hydroxy-6-methyl-2-nitropyridine, pubchem6630, 2-nitro-6-methylpyridin-3-ol, 3-pyridinol,6-methyl-2-nitro, 6-methyl-2-nitro-pyridin-3-ol, wzmgqhibxuaygs-uhfffaoysa | |
| CC1=NC(=C(C=C1)O)[N+](=O)[O-] | |
| 84801 | |
| ≥98.0% (GC) | |
| 3-Hydroxy-6-methyl-2-nitropyridine |
| 107°C | |
| C6H6N2O3 | |
| 5 g | |
| WZMGQHIBXUAYGS-UHFFFAOYSA-N | |
| 154.125 | |
| 154.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : No