missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-6-methyl-2-nitropyridine 98.0+%, TCI America™

Catalog No. H07625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07625G 5 g
H076225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07625G Supplier TCI America Supplier No. H07625G

Chemical Identifiers

CAS 15128-90-2
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00006259
InChI Key WZMGQHIBXUAYGS-UHFFFAOYSA-N
Synonym 3-hydroxy-6-methyl-2-nitropyridine, 6-methyl-2-nitropyridin-3-ol, 6-methyl-2-nitro-3-pyridinol, 3-pyridinol, 6-methyl-2-nitro, 3-hydroxy-2-nitro-6-picoline 3-hydroxy-6-methyl-2-nitropyridine, pubchem6630, 2-nitro-6-methylpyridin-3-ol, 3-pyridinol,6-methyl-2-nitro, 6-methyl-2-nitro-pyridin-3-ol, wzmgqhibxuaygs-uhfffaoysa
PubChem CID 84801
SMILES CC1=NC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 107°C
Color Yellow
Quantity 5 g
Formula Weight 154.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-6-methyl-2-nitropyridine