missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-6-methoxyflavone 98.0+%, TCI America™
$165.90 - $533.13
Chemical Identifiers
| CAS | 93176-00-2 |
|---|---|
| Molecular Formula | C16H12O4 |
| Molecular Weight (g/mol) | 268.268 |
| MDL Number | MFCD00017639 |
| InChI Key | OGURJSOPVFCIOO-UHFFFAOYSA-N |
| Synonym | 3-hydroxy-6-methoxyflavone, 6-methoxyflavonol, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-one, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-on, 4h-1-benzopyran-4-one, 3-hydroxy-6-methoxy-2-phenyl, 3-hydroxy-6-methoxy-2-phenyl-4h-1-benzopyran-4-one |
| PubChem CID | 688676 |
| IUPAC Name | 3-hydroxy-6-methoxy-2-phenylchromen-4-one |
| SMILES | COC1=CC2=C(C=C1)OC(=C(C2=O)O)C3=CC=CC=C3 |
Chemical Identifiers
| 93176-00-2 | |
| 268.268 | |
| OGURJSOPVFCIOO-UHFFFAOYSA-N | |
| 688676 | |
| COC1=CC2=C(C=C1)OC(=C(C2=O)O)C3=CC=CC=C3 |
| C16H12O4 | |
| MFCD00017639 | |
| 3-hydroxy-6-methoxyflavone, 6-methoxyflavonol, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-one, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-on, 4h-1-benzopyran-4-one, 3-hydroxy-6-methoxy-2-phenyl, 3-hydroxy-6-methoxy-2-phenyl-4h-1-benzopyran-4-one | |
| 3-hydroxy-6-methoxy-2-phenylchromen-4-one |
Specifications
| 93176-00-2 | |
| White-Yellow | |
| MFCD00017639 | |
| 3-hydroxy-6-methoxyflavone, 6-methoxyflavonol, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-one, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-on, 4h-1-benzopyran-4-one, 3-hydroxy-6-methoxy-2-phenyl, 3-hydroxy-6-methoxy-2-phenyl-4h-1-benzopyran-4-one | |
| COC1=CC2=C(C=C1)OC(=C(C2=O)O)C3=CC=CC=C3 | |
| 268.268 | |
| 268.27 | |
| Crystalline Powder |
| 208°C | |
| C16H12O4 | |
| 1 g | |
| OGURJSOPVFCIOO-UHFFFAOYSA-N | |
| 3-hydroxy-6-methoxy-2-phenylchromen-4-one | |
| 688676 | |
| ≥98.0% (GC) | |
| 3-Hydroxy-6-methoxyflavone |
Safety and Handling
TSCA : No