missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-5-methylpyridine, 97%

Catalog No. AAH3265806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3265806 5 g
AAH3265803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3265806 Supplier Thermo Scientific Chemicals Supplier No. H3265806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42732-49-0 | C6H7NO | 109.128 g/mol

3-Hydroxy-5-methylpyridine is used to prepare 3-hydroxy-5-methyl-2-nitropyridine by nitration reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-5-methylpyridine is used to prepare 3-hydroxy-5-methyl-2-nitropyridine by nitration reaction.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

Chemical Identifiers

CAS 42732-49-0
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00661297
InChI Key RYJNCIGFPWGVPA-UHFFFAOYSA-N
Synonym 3-hydroxy-5-picoline, 3-hydroxy-5-methylpyridine, 5-methyl-3-hydroxypyridine, 5-hydroxy-3-picoline, 5-methyl-3-pyridinol, 5-methyl-3-pyridol, pubchem6629, 5-methyl-3-hydroxy-pyridine, 3-pyridinol, 5-methyl, 3-hydroxy-5-methylpyridine 250mg
PubChem CID 224753
IUPAC Name 5-methylpyridin-3-ol
SMILES CC1=CC(=CN=C1)O

Specifications

Melting Point 136°C to 140°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-5-methylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only