missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-quinolinecarboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 118-13-8
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD01536959
InChI Key MXNVEJDRXSFZQB-UHFFFAOYSA-N
Synonym 3-hydroxy-4-quinolinecarboxylic acid, 3-hydroxycinchoninic acid, cinchoninic acid, 3-hydroxy, 3-hydroxy-quinoline-4-carboxylic acid, 4-quinolinecarboxylic acid, 3-hydroxy, 3-hydroxy-4-quinolinecarboxylicacid, 3-hydroxycinchoninicacid, 4-22-00-02296 beilstein handbook reference, 4-quinolinecarboxylicacid,3-hydroxy, 4-quinolinecarboxylicacid, 3-hydroxy
PubChem CID 8352
IUPAC Name 3-hydroxyquinoline-4-carboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=NC=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC449930010
SDS View Documents
Thermo Scientific Chemicals
449930010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC449930050
SDS View Documents
Thermo Scientific Chemicals
449930050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

118-13-8
189.17
MXNVEJDRXSFZQB-UHFFFAOYSA-N
8352
OC(=O)C1=C2C=CC=CC2=NC=C1O
C10H7NO3
MFCD01536959
3-hydroxy-4-quinolinecarboxylic acid, 3-hydroxycinchoninic acid, cinchoninic acid, 3-hydroxy, 3-hydroxy-quinoline-4-carboxylic acid, 4-quinolinecarboxylic acid, 3-hydroxy, 3-hydroxy-4-quinolinecarboxylicacid, 3-hydroxycinchoninicacid, 4-22-00-02296 beilstein handbook reference, 4-quinolinecarboxylicacid,3-hydroxy, 4-quinolinecarboxylicacid, 3-hydroxy
3-hydroxyquinoline-4-carboxylic acid

Specifications

118-13-8
Glass Bottle
MFCD01536959
3-hydroxy-4-quinolinecarboxylic acid, 3-hydroxycinchoninic acid, cinchoninic acid, 3-hydroxy, 3-hydroxy-quinoline-4-carboxylic acid, 4-quinolinecarboxylic acid, 3-hydroxy, 3-hydroxy-4-quinolinecarboxylicacid, 3-hydroxycinchoninicacid, 4-22-00-02296 beilstein handbook reference, 4-quinolinecarboxylicacid,3-hydroxy, 4-quinolinecarboxylicacid, 3-hydroxy
OC(=O)C1=C2C=CC=CC2=NC=C1O
189.17
189.17
3-Hydroxy-4-quinolinecarboxylic acid
96% min. (HPLC)
C10H7NO3
1g
MXNVEJDRXSFZQB-UHFFFAOYSA-N
3-hydroxyquinoline-4-carboxylic acid
8352
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 204-236-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only