missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4'-methoxyflavone 98.0+%, TCI America™

Catalog No. H14051G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14051G 1 g
H14055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14051G Supplier TCI America Supplier No. H14051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6889-78-7
Molecular Formula C16H12O4
Molecular Weight (g/mol) 268.268
MDL Number MFCD00017682
InChI Key IIBBFGMVMNZMGA-UHFFFAOYSA-N
Synonym 4'-methoxyflavonol, 3-hydroxy-4'-methoxyflavone, 3-hydroxy-2-4-methoxyphenyl-4h-chromen-4-one, 3-hydroxy-2-4-methoxyphenyl chromen-4-one, 4h-1-benzopyran-4-one, 3-hydroxy-2-4-methoxyphenyl, 2-4-methoxyphenyl-3-hydroxyflavone, hydroxymethoxyphenylchromenone, 3-hydroxy-4'-methoxyflavanone, 4h-1-benzopyran-4-one, 3-hydroxy-2-4-methoxyphenyl-9ci
PubChem CID 97141
IUPAC Name 3-hydroxy-2-(4-methoxyphenyl)chromen-4-one
SMILES COC1=CC=C(C=C1)C2=C(C(=O)C3=CC=CC=C3O2)O

Specifications

Melting Point 234°C
Color Yellow
Quantity 1 g
Formula Weight 268.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4′-methoxyflavone