missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-4-methoxycinnamic Acid 98.0+%, TCI America™
$104.38 - $410.10
Chemical Identifiers
| CAS | 537-73-5 |
|---|---|
| Molecular Formula | C10H10O4 |
| Molecular Weight (g/mol) | 194.186 |
| MDL Number | MFCD00004391 |
| InChI Key | QURCVMIEKCOAJU-HWKANZROSA-N |
| Synonym | isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid |
| PubChem CID | 736186 |
| ChEBI | CHEBI:27794 |
| IUPAC Name | (E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid |
| SMILES | COC1=C(C=C(C=C1)C=CC(=O)O)O |
Chemical Identifiers
| 537-73-5 | |
| 194.186 | |
| QURCVMIEKCOAJU-HWKANZROSA-N | |
| 736186 | |
| (E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid |
| C10H10O4 | |
| MFCD00004391 | |
| isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid | |
| CHEBI:27794 | |
| COC1=C(C=C(C=C1)C=CC(=O)O)O |
Specifications
| 537-73-5 | |
| White-Yellow | |
| MFCD00004391 | |
| isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid | |
| COC1=C(C=C(C=C1)C=CC(=O)O)O | |
| 194.186 | |
| CHEBI:27794 | |
| ≥98.0% (HPLC,T) | |
| 3-Hydroxy-4-methoxycinnamic Acid |
| 228°C | |
| C10H10O4 | |
| 1 g | |
| QURCVMIEKCOAJU-HWKANZROSA-N | |
| (E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid | |
| 736186 | |
| 194.19 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UD3365400
TSCA : No