missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxycinnamic Acid 98.0+%, TCI America™

Catalog No. H05241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05241G 1 g
H05245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05241G Supplier TCI America Supplier No. H05241G

Chemical Identifiers

CAS 537-73-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004391
InChI Key QURCVMIEKCOAJU-HWKANZROSA-N
Synonym isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid
PubChem CID 736186
ChEBI CHEBI:27794
IUPAC Name (E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=C(C=C1)C=CC(=O)O)O

Specifications

Melting Point 228°C
Color White-Yellow
Quantity 1 g
Formula Weight 194.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4-methoxycinnamic Acid
RTECSNumber UD3365400
TSCA No