missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 98+%

CAS: 537-73-5 | C10H10O4 | 194.186 g/mol

$72.74 - $270.10

Chemical Identifiers

CAS 537-73-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004391
InChI Key QURCVMIEKCOAJU-HWKANZROSA-N
Synonym isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid
PubChem CID 736186
ChEBI CHEBI:27794
IUPAC Name (E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=C(C=C1)C=CC(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1048203
View Documents
Thermo Scientific Chemicals
A1048203
1 g
Each for $72.74
Only null left
 
AAA1048206
View Documents
Thermo Scientific Chemicals
A1048206
5 g
Each for $270.10
Only null left
 
Description

Description

3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-4-methoxycinnamic acid exhibits hypoglycemic properties.

Solubility
Soluble in ethanol and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under normal temperatures and pressures.
Specifications

Chemical Identifiers

537-73-5
194.186
QURCVMIEKCOAJU-HWKANZROSA-N
736186
(E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
C10H10O4
MFCD00004391
isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid
CHEBI:27794
COC1=C(C=C(C=C1)C=CC(=O)O)O

Specifications

537-73-5
C10H10O4
1 g
isoferulic acid, 3-hydroxy-4-methoxycinnamic acid, hesperetic acid, 3-3-hydroxy-4-methoxyphenyl acrylic acid, 2e-3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, isoferulate, 2-propenoic acid, 3-3-hydroxy-4-methoxyphenyl, e-3-3-hydroxy-4-methoxyphenyl acrylic acid, 3-3-hydroxy-4-methoxyphenyl prop-2-enoic acid, 3-hydroxy-4-methoxy-cinnamic acid
QURCVMIEKCOAJU-HWKANZROSA-N
(E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
736186
194.19
3-Hydroxy-4-methoxycinnamic acid, predominantly trans
∼231°C (decomposition)
MFCD00004391
2212760
Soluble in ethanol and methanol.
COC1=C(C=C(C=C1)C=CC(=O)O)O
194.186
CHEBI:27794
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-676-0

RTECSNumber : UD3365400

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only