missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxybenzaldehyde, 98%

Catalog No. AC162571000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162571000 100 g Plastic bottle
AC162570050 5 g Glass bottle
AC162570250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC162571000 Supplier Thermo Scientific Chemicals Supplier No. 162571000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 621-59-0 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-59-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003369
InChI Key JVTZFYYHCGSXJV-UHFFFAOYSA-N
Synonym isovanillin, isovanilline, 3-hydroxy-p-anisaldehyde, 5-formylguaiacol, benzaldehyde, 3-hydroxy-4-methoxy, iso-vanillin, 3-hydroxyanisaldehyde, isovanilin, isovanicaline, 3-hydroxy-4-methoxy-benzaldehyde
PubChem CID 12127
IUPAC Name 3-hydroxy-4-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 112°C to 116°C
Color Brown or Gray to White
Boiling Point 179°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula HOC6H3(OCH3)CHO
Quantity 100 g
Beilstein 08, 254
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Hydroxy-4-methoxybenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-694-9

RUO – Research Use Only