missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxybenzaldehyde, 98%

Catalog No. AAA1286606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1286606 5 g
AAA1286614 25 g
AAA1286622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1286606 Supplier Thermo Scientific Chemicals Supplier No. A1286606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 621-59-0 | C8H8O3 | 152.149 g/mol

3-Hydroxy-4-methoxybenzaldehyde acts as a precursor for the stereoselective synthesis of the anticancer drug (Z)-combretastatin A-4 and glycitein. It also used as an important raw material for the preparation of morphine. Further, it is involved in the preparation of Schiff- bases by reacting with furan-2-carboxylic acid hydrazide and thiophene-2-carboxylic acid hydrazide. In addition to this, it is used to prepare (Z)-2-(3-hydroxy-4-methoxybenzylidene)-1-azabicyclo[2.2.2]octan-3-one by reacting with with1-azabicyclo[2.2.2]octan-3-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-4-methoxybenzaldehyde acts as a precursor for the stereoselective synthesis of the anticancer drug (Z)-combretastatin A-4 and glycitein. It also used as an important raw material for the preparation of morphine. Further, it is involved in the preparation of Schiff- bases by reacting with furan-2-carboxylic acid hydrazide and thiophene-2-carboxylic acid hydrazide. In addition to this, it is used to prepare (Z)-2-(3-hydroxy-4-methoxybenzylidene)-1-azabicyclo[2.2.2]octan-3-one by reacting with with1-azabicyclo[2.2.2]octan-3-one.

Solubility
Soluble in acetone and methanol.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 621-59-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00003369
InChI Key JVTZFYYHCGSXJV-UHFFFAOYSA-N
Synonym isovanillin, isovanilline, 3-hydroxy-p-anisaldehyde, 5-formylguaiacol, benzaldehyde, 3-hydroxy-4-methoxy, iso-vanillin, 3-hydroxyanisaldehyde, isovanilin, isovanicaline, 3-hydroxy-4-methoxy-benzaldehyde
PubChem CID 12127
IUPAC Name 3-hydroxy-4-methoxybenzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)O

Specifications

Melting Point 112°C to 116°C
Density 1.2
Boiling Point 179°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1073021
Sensitivity Air and light sensitive
Solubility Information Soluble in acetone and methanol.
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-4-methoxybenzaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-694-9
RTECSNumber CU6540000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only