missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4,5-dimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. H11795G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11795G 5 g
H11791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11795G Supplier TCI America Supplier No. H11795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29865-90-5
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00010442
InChI Key NVLTWXMZECWWPC-UHFFFAOYSA-N
Synonym 5-Hydroxyveratraldehyde, 5-Hydroxy-3,4-dimethoxybenzaldehyde
PubChem CID 141513
IUPAC Name 3-hydroxy-4,5-dimethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(O)=C1OC

Specifications

Melting Point 66°C
Boiling Point 180°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4,5-dimethoxybenzaldehyde