missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-pyrrolidinone, 95%, Thermo Scientific Chemicals

Catalog No. aah5796103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5796103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5796103 Supplier Thermo Scientific Chemicals Supplier No. H5796103
Only null left

3-Hydroxy-2-pyrrolidinone is used as a potent inhibitor of CDK2/cyclin A. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-2-pyrrolidinone is used as a potent inhibitor of CDK2/cyclin A. It is also used as pharmaceutical intermediates.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Store under dry inert gas. Incompatible with heat, oxidizing agents and moisture.

Chemical Identifiers

CAS 15166-68-4
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD09751256
InChI Key FRKGSNOMLIYPSH-UHFFFAOYSA-N
Synonym 3-hydroxy-2-pyrrolidinone, 2-pyrrolidinone, 3-hydroxy, 3-hydroxy-pyrrolidin-2-one, a-hydroxy-?-butyrolactam, acmc-209pe0, 3-oxidanylpyrrolidin-2-one, 3-hydroxy-2-oxopyrrolidine, 3-hydroxypyrrolidine-2-one
PubChem CID 10240772
IUPAC Name 3-hydroxypyrrolidin-2-one
SMILES C1CNC(=O)C1O

Specifications

Melting Point 102°C to 107°C
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 101.11
Percent Purity 95%
Chemical Name or Material 3-Hydroxy-2-pyrrolidinone
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only