missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-nitropyridine, 98%

CAS: 15128-82-2 | C5H4N2O3 | 140.10 g/mol

Supplier:  Thermo Scientific Chemicals A1488730

 View more versions of this product

Catalog No. AAA1488730


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Hydroxy-2-nitropyridine
69°C to 73°C
MFCD00006258
124473
QBPDSKPWYWIHGA-UHFFFAOYSA-N
2-nitropyridin-3-ol
27057
98%
15128-82-2
C5H4N2O3
250 g
3-hydroxy-2-nitropyridine, 2-nitropyridin-3-ol, 2-nitro-3-hydroxypyridine, 2-nitro-3-pyridinol, 3-pyridinol, 2-nitro, 2-nitro-3-hydroxy pyridine, unii-uu357x85yb, 3-hydroxy-2-nitro pyridine, 2-nitro-3-pyridol
OC1=CC=CN=C1[N+]([O-])=O
140.10
140.1

Chemical Identifiers

15128-82-2
140.10
QBPDSKPWYWIHGA-UHFFFAOYSA-N
27057
OC1=CC=CN=C1[N+]([O-])=O
C5H4N2O3
MFCD00006258
3-hydroxy-2-nitropyridine, 2-nitropyridin-3-ol, 2-nitro-3-hydroxypyridine, 2-nitro-3-pyridinol, 3-pyridinol, 2-nitro, 2-nitro-3-hydroxy pyridine, unii-uu357x85yb, 3-hydroxy-2-nitro pyridine, 2-nitro-3-pyridol
2-nitropyridin-3-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 239-191-2

RTECSNumber : UU7715000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only