missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-2-methylpyridine 98.0+%, TCI America™
$192.00 - $628.19
Chemical Identifiers
| CAS | 1121-25-1 |
|---|---|
| Molecular Formula | C6H7NO |
| Molecular Weight (g/mol) | 109.128 |
| MDL Number | MFCD00082538 |
| InChI Key | AQSRRZGQRFFFGS-UHFFFAOYSA-N |
| Synonym | 3-hydroxy-2-methylpyridine, 2-methyl-3-hydroxypyridine, 3-hydroxy-2-picoline, 2-methyl-3-pyridinol, 3-pyridinol, 2-methyl, 2-methyl-3-hydroxylpyridine, unii-365fj23a8z, 3-pyridinol, methyl, 2-methyl-pyridin-3-ol, 3-hydroxy-2-methylpyridine 3-hydroxy-2-picoline |
| PubChem CID | 70719 |
| IUPAC Name | 2-methylpyridin-3-ol |
| SMILES | CC1=C(C=CC=N1)O |
Chemical Identifiers
| 1121-25-1 | |
| 109.128 | |
| AQSRRZGQRFFFGS-UHFFFAOYSA-N | |
| 70719 | |
| CC1=C(C=CC=N1)O |
| C6H7NO | |
| MFCD00082538 | |
| 3-hydroxy-2-methylpyridine, 2-methyl-3-hydroxypyridine, 3-hydroxy-2-picoline, 2-methyl-3-pyridinol, 3-pyridinol, 2-methyl, 2-methyl-3-hydroxylpyridine, unii-365fj23a8z, 3-pyridinol, methyl, 2-methyl-pyridin-3-ol, 3-hydroxy-2-methylpyridine 3-hydroxy-2-picoline | |
| 2-methylpyridin-3-ol |
Specifications
| 1121-25-1 | |
| C6H7NO | |
| 1 g | |
| AQSRRZGQRFFFGS-UHFFFAOYSA-N | |
| 2-methylpyridin-3-ol | |
| 70719 | |
| ≥98.0% (GC,T) | |
| 3-Hydroxy-2-methylpyridine |
| 169°C | |
| MFCD00082538 | |
| 3-hydroxy-2-methylpyridine, 2-methyl-3-hydroxypyridine, 3-hydroxy-2-picoline, 2-methyl-3-pyridinol, 3-pyridinol, 2-methyl, 2-methyl-3-hydroxylpyridine, unii-365fj23a8z, 3-pyridinol, methyl, 2-methyl-pyridin-3-ol, 3-hydroxy-2-methylpyridine 3-hydroxy-2-picoline | |
| CC1=C(C=CC=N1)O | |
| 109.128 | |
| 109.13 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3689
TSCA : Yes