missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methylpyridine 98.0+%, TCI America™

Catalog No. H13655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13655G 5 g
H13651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13655G Supplier TCI America Supplier No. H13655G

Chemical Identifiers

CAS 1121-25-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00082538
InChI Key AQSRRZGQRFFFGS-UHFFFAOYSA-N
Synonym 3-hydroxy-2-methylpyridine, 2-methyl-3-hydroxypyridine, 3-hydroxy-2-picoline, 2-methyl-3-pyridinol, 3-pyridinol, 2-methyl, 2-methyl-3-hydroxylpyridine, unii-365fj23a8z, 3-pyridinol, methyl, 2-methyl-pyridin-3-ol, 3-hydroxy-2-methylpyridine 3-hydroxy-2-picoline
PubChem CID 70719
IUPAC Name 2-methylpyridin-3-ol
SMILES CC1=C(C=CC=N1)O

Specifications

Melting Point 169°C
Quantity 5 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-methylpyridine
EINECSNumber (5)-3689
TSCA Yes