missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methylpyridine, 99%

Catalog No. AAB2494414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2494414 25 g
AAB2494403 1 g
AAB2494406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2494414 Supplier Thermo Scientific Chemicals Supplier No. B2494414

CAS: 1121-25-1 | C6H7NO | 109.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1121-25-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00082538
InChI Key AQSRRZGQRFFFGS-UHFFFAOYSA-N
Synonym 3-hydroxy-2-methylpyridine, 2-methyl-3-hydroxypyridine, 3-hydroxy-2-picoline, 2-methyl-3-pyridinol, 3-pyridinol, 2-methyl, 2-methyl-3-hydroxylpyridine, unii-365fj23a8z, 3-pyridinol, methyl, 2-methyl-pyridin-3-ol, 3-hydroxy-2-methylpyridine 3-hydroxy-2-picoline
PubChem CID 70719
IUPAC Name 2-methylpyridin-3-ol
SMILES CC1=C(C=CC=N1)O

Specifications

Melting Point 166°C to 169°C
Odor Odorless
Quantity 25 g
Beilstein 107937
Formula Weight 109.13
Percent Purity 99%
Chemical Name or Material 3-Hydroxy-2-methylpyridine
Hazard Category H302-H315-H318-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 214-327-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only