missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methyl-4-pyrone 99.0+%, TCI America™

Catalog No. M0673500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0673500G 500 g
M067325G 25 g
M0673100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0673500G Supplier TCI America Supplier No. M0673500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118-71-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006578
InChI Key XPCTZQVDEJYUGT-UHFFFAOYSA-N
Synonym maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl
PubChem CID 8369
ChEBI CHEBI:69438
IUPAC Name 3-hydroxy-2-methyl-4H-pyran-4-one
SMILES CC1=C(O)C(=O)C=CO1

Specifications

Melting Point 162°C
Color White
Quantity 500 g
Formula Weight 126.11
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-methyl-4-pyrone
EINECSNumber (5)-3642
RTECSNumber UQ1050000
TSCA Yes