missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-methyl-4-pyrone, 99%

Catalog No. p-7080967
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1073514 25 g
AAA1073522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1073514 Supplier Thermo Scientific Chemicals Supplier No. A1073514

CAS: 118-71-8 | C6H6O3 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-71-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006578
InChI Key XPCTZQVDEJYUGT-UHFFFAOYSA-N
Synonym maltol, 3-hydroxy-2-methyl-4-pyrone, 3-hydroxy-2-methyl-4h-pyran-4-one, larixinic acid, talmon, palatone, vetol, larixic acid, corps praline, 4h-pyran-4-one, 3-hydroxy-2-methyl
PubChem CID 8369
ChEBI CHEBI:69438
IUPAC Name 3-hydroxy-2-methylpyran-4-one
SMILES CC1=C(O)C(=O)C=CO1

Specifications

Melting Point 161°C to 165°C
Boiling Point 170°C (100 mmHg)
Flash Point 95°C (203°F)
Quantity 25 g
Beilstein 112169
Merck Index 14,5713
Formula Weight 126.11
Percent Purity 99%
Chemical Name or Material 3-Hydroxy-2-methyl-4-pyrone
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-271-8
RTECSNumber UQ1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only