missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-butanone, monomer + dimer, 95%

Catalog No. AAA1375218
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1375218 50 g
AAA1375230 250 g
AAA137520B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1375218 Supplier Thermo Scientific Chemicals Supplier No. A1375218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 513-86-0 | C4H8O2 | 88.11 g/mol

3-Hydroxy-2-butanone is a chemical used in food flavoring and fragrances. It acts as an intermediate of butanediol cycle in microorganisms. It is used as an aroma carrier in the preparation of flavors and essences.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxy-2-butanone is a chemical used in food flavoring and fragrances. It acts as an intermediate of butanediol cycle in microorganisms. It is used as an aroma carrier in the preparation of flavors and essences.

Solubility
Miscible with water, propylene glycol and alcohol. Slightly miscible with ether, petroleum ether. Immiscible with fatty oils and vegetable oil.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 513-86-0
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00004521,MFCD00038696
InChI Key ROWKJAVDOGWPAT-UHFFFAOYNA-N
Synonym acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl
PubChem CID 179
ChEBI CHEBI:15688
IUPAC Name 3-hydroxybutan-2-one
SMILES CC(O)C(C)=O

Specifications

Density 1.013
Boiling Point 148°C
Flash Point 41°C (106°F)
Odor Characteristic
Quantity 50 g
UN Number UN2621
Beilstein 385636
Merck Index 14,64
Solubility Information Miscible with water,propylene glycol and alcohol. Slightly miscible with ether,petroleum ether. Immiscible with fatty oils and vegetable oil.
Formula Weight 88.11
Percent Purity 95%
Chemical Name or Material 3-Hydroxy-2-butanone, monomer + dimer
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ACETYL METHYL CARBINOL
EINECSNumber 208-174-1
RTECSNumber EL8790000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only